Search
Search

CaS| Gene Symbol: CaS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122212.5 | NP_197697.1 | 5 - 6 | 1052 | 123 | |
| GenBank mRNA | AY045798.1 | - | 5 - 6 | 1027 | 123 | |
| AY079331.1 | - | 5 - 6 | 929 | 123 | ||
| AY341888.1 | - | 5 - 6 | 1014 | 123 | ||
| BT000666.1 | - | 5 - 6 | 1031 | 123 | ||
Set Membership: |
|