Search
Search

AGL87| Gene Symbol: AGL87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102106.4 | NP_564174.1 | 2 - 3 | 220 | 87 | |
| GenBank mRNA | AK117835.1 | - | 2 - 3 | 152 | 87 | |
| BT043486.1 | - | 2 - 3 | 93 | 87 | ||
Set Membership: |
|