Search
Search

ATSAC1| Gene Symbol: ATSAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102109.4 | NP_173676.2 | 5 - 6 | 1005 | 129 | |
| GenBank mRNA | AF266460.1 | - | 5 - 6 | 803 | 129 | |
| AY227244.1 | - | 5 - 6 | 803 | 129 | ||
Set Membership: |
|