Search
Search

AT1G22850| Gene Symbol: AT1G22850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332560.1 | NP_001322655.1 | 4 - 5 | 874 | 63 | |
| NM_102131.5 | NP_564182.1 | 4 - 5 | 881 | 63 | ||
| GenBank mRNA | AY050329.1 | - | 4 - 5 | 836 | 63 | |
| AY101531.1 | - | 4 - 5 | 815 | 63 | ||
Set Membership: |
|