Search
Search

AT1G26940| Gene Symbol: AT1G26940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332736.1 | NP_001320716.1 | 7 - 8 | 791 | 140 | |
| NM_102458.2 | NP_564267.1 | 7 - 8 | 738 | 140 | ||
| GenBank mRNA | AK118273.1 | - | 7 - 8 | 712 | 140 | |
| AY085848.1 | - | 7 - 8 | 726 | 140 | ||
Set Membership: |
|