Search
Search

AT1G28120| Gene Symbol: AT1G28120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102577.4 | NP_564299.1 | 7 - 8 | 734 | 91 | |
| GenBank mRNA | AY136373.1 | - | 7 - 8 | 717 | 91 | |
| BT000164.1 | - | 6 - 7 | 519 | 91 | ||
| JQ013442.1 | - | 6 - 7 | 519 | 91 | ||
Set Membership: |
|