Search
Search

ATR| Gene Symbol: ATR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344355.1 | NP_001332644.1 | 6 - 7 | 1074 | 131 | |
| NM_001344356.1 | NP_001332643.1 | 6 - 7 | 1074 | 131 | ||
| NM_001344357.1 | NP_001332642.1 | 6 - 7 | 1074 | 131 | ||
| NM_123447.3 | NP_198898.2 | 6 - 7 | 1074 | 131 | ||
Set Membership: |
|