Search
Search

UMAMIT7| Gene Symbol: UMAMIT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344725.1 | NP_001330912.1 | 4 - 5 | 845 | 64 | |
| NM_001344726.1 | NP_001330913.1 | 4 - 5 | 821 | 64 | ||
| NM_124120.1 | NP_199558.1 | 4 - 5 | 650 | 64 | ||
Set Membership: |
|