Search
Search

CLC-C| Gene Symbol: CLC-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124367.4 | NP_199800.1 | 5 - 6 | 1407 | 91 | |
| GenBank mRNA | AY120754.1 | - | 5 - 6 | 1238 | 91 | |
| BT001211.1 | - | 5 - 6 | 1118 | 91 | ||
| Y09095.1 | - | 1141 | 91 | |||
| Z71447.1 | - | 5 - 6 | 1151 | 91 | ||
Set Membership: |
|