Search
Search

GPX3| Gene Symbol: GPX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202813.1 | NP_001189742.1 | 4 - 5 | 496 | 134 | |
| NM_129896.4 | NP_181863.1 | 4 - 5 | 504 | 134 | ||
| GenBank mRNA | AY065372.1 | - | 4 - 5 | 491 | 134 | |
| AY087030.1 | - | 4 - 5 | 489 | 134 | ||
| AY096479.1 | - | 4 - 5 | 423 | 134 | ||
Set Membership: |
|