Search
Search

AT4G30410| Gene Symbol: AT4G30410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119187.4 | NP_194770.1 | 2 | 836 | 89 | |
| NM_202911.2 | NP_974640.1 | 1 | 1036 | 89 | ||
| GenBank mRNA | AK227688.1 | - | 1 | 946 | 89 | |
Set Membership: |
|