Search
Search

WRKY35| Gene Symbol: WRKY35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336530.1 | NP_001324223.1 | 1 - 2 | 991 | 151 | |
| NM_129036.2 | NP_181029.1 | 1 - 2 | 566 | 151 | ||
| GenBank mRNA | AF404863.1 | - | 1 - 2 | 566 | 151 | |
| BT029239.1 | - | 1 - 2 | 566 | 151 | ||
Set Membership: |
|