Search
Search

GSTF2| Gene Symbol: GSTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116486.3 | NP_192161.1 | 3 | 1275 | 101 | |
| GenBank mRNA | AF326903.1 | - | 3 | 862 | 101 | |
| AY039580.1 | - | 3 | 862 | 101 | ||
| X75303.1 | - | 815 | 101 | |||
Set Membership: |
|