Search
Search

GTPBP1| Gene Symbol: GTPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017938.1 | NP_001017938.1 | 2 - 3 | 301 | 64 | |
| GenBank mRNA | BC151310.1 | - | 2 - 3 | 320 | 64 | |
| BT021672.1 | - | 2 - 3 | 301 | 64 | ||
Set Membership: |
|