Search
Search
| Gene Symbol: OGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098070.2 | NP_001091539.1 | 14 - 15 | 2021 | 82 | |
| GenBank mRNA | BC140542.1 | - | 14 - 15 | 2022 | 82 | |
| BC149782.1 | - | 10 - 11 | 1932 | 82 | ||
Set Membership: |
|