Search
Search

UBQLN1| Gene Symbol: UBQLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174628.1 | NP_777053.1 | 10 - 11 | 1620 | 68 | |
| GenBank mRNA | AY035552.1 | - | 10 - 11 | 1620 | 68 | |
| BC120031.1 | - | 10 - 11 | 1844 | 68 | ||
Set Membership: |
|