Search
Search

NPC1| Gene Symbol: NPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174758.2 | NP_777183.1 | 6 - 7 | 955 | 107 | |
| GenBank mRNA | AF299073.1 | - | 6 - 7 | 955 | 107 | |
| BC151276.1 | - | 6 - 7 | 993 | 107 | ||
Set Membership: |
|