Search
Search

YWHAZ| Gene Symbol: YWHAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174814.2 | NP_777239.1 | 4 - 5 | 631 | 106 | |
| GenBank mRNA | BC102382.1 | - | 4 - 5 | 665 | 106 | |
| L07955.1 | - | 4 - 5 | 631 | 106 | ||
Set Membership: |
|