Search
Search

PRICKLE3| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014886.1 | NP_001014886.1 | 8 - 9 | 1326 | 66 | |
| GenBank mRNA | BT021566.1 | - | 8 - 9 | 1326 | 66 | |
Set Membership: |
|