Search
Search

RAB13| Gene Symbol: RAB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024540.1 | NP_001019711.1 | 2 - 3 | 300 | 130 | |
| GenBank mRNA | BC109840.1 | - | 2 - 3 | 302 | 130 | |
| BT021522.1 | - | 2 - 3 | 300 | 130 | ||
Set Membership: |
|