Search
Search

BTF3| Gene Symbol: BTF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025314.2 | NP_001020485.1 | 4 - 5 | 501 | 66 | |
| GenBank mRNA | AY911339.1 | - | 4 - 5 | 502 | 66 | |
| BC102975.1 | - | 4 - 5 | 498 | 66 | ||
Set Membership: |
|