Search
Search

ADIPOR1| Gene Symbol: ADIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034055.1 | NP_001029227.1 | 7 - 8 | 1183 | 67 | |
| GenBank mRNA | AY436318.1 | - | 2 - 3 | 219 | 67 | |
| BC102259.1 | - | 7 - 8 | 1183 | 67 | ||
Set Membership: |
|