Search
Search

SPARCL1| Gene Symbol: SPARCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034302.2 | NP_001029474.1 | 9 - 10 | 2028 | 77 | |
| GenBank mRNA | BC103351.1 | - | 9 - 10 | 2029 | 77 | |
Set Membership: |
|