Search
Search

GSTO1| Gene Symbol: GSTO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075214.2 | NP_001068682.2 | 4 - 5 | 546 | 87 | |
| GenBank mRNA | DV852688.1 | - | 2 - 3 | 281 | 87 | |
| EE226879.1 | - | 4 - 5 | 546 | 87 | ||
Set Membership: |
|