Search
Search

AQP8| Gene Symbol: AQP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206607.3 | NP_001193536.1 | 4 - 5 | 712 | 78 | |
| GenBank mRNA | AY743596.1 | - | 2 - 3 | 322 | 78 | |
| BC116016.1 | - | 4 - 5 | 687 | 78 | ||
Set Membership: |
|