Search
Search

AP3B2| Gene Symbol: AP3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243335.1 | NP_001230264.1 | 1 - 2 | 301 | 85 | |
| GenBank mRNA | BC149947.1 | - | 1 - 2 | 301 | 85 | |
| DV906389.1 | - | 1 - 2 | 301 | 85 | ||
Set Membership: |
|