Search
Search

hpo-13| Gene Symbol: hpo-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026510.4 | NP_001021681.1 | 2 - 3 | 267 | 147 | |
| NM_001373663.2 | NP_001359639.1 | 2 - 3 | 245 | 147 | ||
| NM_001383760.1 | 2 - 3 | 289 | 147 | |||
Set Membership: |
|