Search
Search

dpf-6| Gene Symbol: dpf-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382930.1 | 5 - 6 | 1211 | 122 | ||
| NM_171203.6 | NP_741240.1 | 5 - 6 | 1209 | 122 | ||
| NM_171205.6 | NP_741242.1 | 2 - 3 | 516 | 122 | ||
Set Membership: |
|