Search
Search

che-13| Gene Symbol: che-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001264468.3 | NP_001251397.1 | 6 - 7 | 724 | 69 | |
| NM_001264469.3 | NP_001251398.1 | 3 - 4 | 383 | 69 | ||
| GenBank mRNA | AF520567.1 | - | 6 - 7 | 644 | 69 | |
Set Membership: |
|