Search
Search

| Gene Symbol: arl-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037897.3 | NP_001032986.1 | 3 - 4 | 306 | 88 | |
| NM_001037898.3 | NP_001032987.1 | 3 - 4 | 332 | 88 | ||
| NM_001373681.2 | NP_001360352.1 | 3 - 4 | 326 | 88 | ||
| Gene Symbol: moag-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383786.1 | 4 - 5 | 985 | 88 | ||
Set Membership: |
|