Search
Search

gcy-17| Gene Symbol: gcy-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306398.2 | NP_001293327.1 | 2 - 3 | 208 | 72 | |
| NM_001306399.3 | NP_001293328.1 | 2 - 3 | 208 | 72 | ||
Set Membership: |
|