Search
Search

sulp-6| Gene Symbol: sulp-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373791.2 | NP_001360427.1 | 10 - 11 | 2208 | 117 | |
| NM_058737.5 | NP_491138.2 | 10 - 11 | 2191 | 117 | ||
Set Membership: |
|