Search
Search

unc-57| Gene Symbol: unc-57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047215.5 | NP_001040680.1 | 2 - 3 | 473 | 148 | |
| NM_001129040.5 | NP_001122512.1 | 2 - 3 | 475 | 148 | ||
| NM_001129041.3 | NP_001122513.1 | 2 - 3 | 473 | 148 | ||
Set Membership: |
|