Search
Search

chtl-1| Gene Symbol: chtl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380966.2 | NP_001367416.1 | 9 - 10 | 2220 | 66 | |
| NM_001380967.2 | NP_001367417.1 | 9 - 10 | 2228 | 66 | ||
| NM_171686.7 | NP_741790.2 | 11 - 12 | 2122 | 66 | ||
Set Membership: |
|