Search
Search

kqt-1| Gene Symbol: kqt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171709.6 | NP_741819.1 | 12 - 13 | 1368 | 95 | |
| NM_171710.7 | NP_741820.1 | 11 - 12 | 1383 | 95 | ||
| GenBank mRNA | AY572974.1 | - | 11 - 12 | 1214 | 95 | |
Set Membership: |
|