Search
Search

gar-2| Gene Symbol: gar-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027422.4 | NP_001022593.1 | 10 - 11 | 1600 | 98 | |
| GenBank mRNA | AY053365.1 | - | 10 - 11 | 1557 | 98 | |
Set Membership: |
|