Search
Search

| Gene Symbol: egl-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313583.2 | NP_001300512.1 | 1 - 2 | 214 | 92 | |
| NM_061251.5 | NP_493652.1 | 1 - 2 | 214 | 92 | ||
| Gene Symbol: C50D2.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313595.3 | NP_001300524.1 | 3 | 216 | 92 | |
Set Membership: |
|