Search
Search

gcy-19| Gene Symbol: gcy-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361852.3 | NP_001348701.1 | 14 | 2323 | 67 | |
| NM_001361853.3 | NP_001348700.1 | 14 - 15 | 2393 | 67 | ||
| NM_001361854.3 | NP_001348702.1 | 15 | 2356 | 67 | ||
Set Membership: |
|