Search
Search

glb-9| Gene Symbol: glb-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001368597.3 | NP_001355411.1 | 6 - 7 | 726 | 121 | |
| NM_001383889.1 | 5 - 6 | 720 | 121 | |||
| NM_063171.6 | NP_495572.3 | 7 - 8 | 816 | 121 | ||
| GenBank mRNA | EF471980.1 | - | 5 - 6 | 636 | 121 | |
Set Membership: |
|