Search
Search

ufd-2| Gene Symbol: ufd-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027149.5 | NP_001022320.1 | 2 - 3 | 382 | 106 | |
| NM_001383900.1 | 2 - 3 | 380 | 106 | |||
| NM_063290.3 | NP_495691.1 | 2 - 3 | 394 | 106 | ||
Set Membership: |
|