Search
Search

mel-11| Gene Symbol: mel-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026757.4 | NP_001021928.1 | 14 - 15 | 3123 | 64 | |
| NM_001381562.1 | NP_001367038.1 | 13 - 14 | 2813 | 64 | ||
| NM_001383917.1 | 12 - 13 | 2660 | 64 | |||
| NM_063593.6 | NP_495994.1 | 13 - 14 | 2805 | 64 | ||
| GenBank mRNA | U86640.1 | - | 13 - 14 | 2796 | 64 | |
Set Membership: |
|