Search
Search

clh-2| Gene Symbol: clh-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026729.4 | NP_001021900.1 | 8 - 9 | 1105 | 129 | |
| NM_001377875.3 | NP_001364585.1 | 8 - 9 | 1183 | 129 | ||
| NM_001383953.1 | 7 - 8 | 993 | 129 | |||
| GenBank mRNA | AF173171.1 | - | 7 - 8 | 1015 | 129 | |
Set Membership: |
|