Search
Search

ceh-100| Gene Symbol: ceh-100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027257.6 | NP_001022428.1 | 10 - 11 | 1735 | 57 | |
| NM_001027258.5 | NP_001022429.1 | 10 - 11 | 1741 | 57 | ||
Set Membership: |
|