Search
Search

tbc-17| Gene Symbol: tbc-17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027263.7 | NP_001022434.2 | 8 - 9 | 1612 | 88 | |
| NM_001027264.7 | NP_001022435.2 | 8 - 9 | 1614 | 88 | ||
| NM_001267427.4 | NP_001254356.1 | 2 - 3 | 718 | 88 | ||
Set Membership: |
|