Search
Search

unc-79| Gene Symbol: unc-79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267957.3 | NP_001254886.1 | 20 - 21 | 5120 | 71 | |
| NM_001381816.2 | NP_001366699.1 | 19 - 20 | 5054 | 71 | ||
Set Membership: |
|