Search
Search

gly-6| Gene Symbol: gly-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027473.4 | NP_001022644.1 | 9 - 10 | 1862 | 122 | |
| NM_001267984.4 | NP_001254913.1 | 1 - 2 | 109 | 122 | ||
| GenBank mRNA | AF031838.1 | - | 8 - 9 | 1593 | 122 | |
Set Membership: |
|