Search
Search

pxl-1| Gene Symbol: pxl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026014.4 | NP_001021185.2 | 6 - 7 | 558 | 64 | |
| NM_001026015.3 | NP_001021186.1 | 2 - 3 | 356 | 64 | ||
| NM_001129205.3 | NP_001122677.1 | 4 - 5 | 381 | 64 | ||
| GenBank mRNA | EU239658.1 | - | 6 - 7 | 553 | 64 | |
| EU239659.1 | - | 2 - 3 | 292 | 64 | ||
| EU239660.1 | - | 4 - 5 | 370 | 64 | ||
Set Membership: |
|