Search
Search

dct-6| Gene Symbol: dct-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038268.4 | NP_001033357.2 | 11 - 12 | 2461 | 71 | |
| NM_001038269.5 | NP_001033358.2 | 12 - 13 | 2485 | 71 | ||
Set Membership: |
|