Search
Search

glr-2| Gene Symbol: glr-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025942.4 | NP_001021113.1 | 6 - 7 | 679 | 116 | |
| NM_001025943.3 | NP_001021114.1 | 6 - 7 | 724 | 116 | ||
| GenBank mRNA | AF318606.2 | - | 6 - 7 | 597 | 116 | |
Set Membership: |
|