Search
Search

spat-2| Gene Symbol: spat-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347290.2 | NP_001334218.1 | 3 - 4 | 373 | 84 | |
| NM_001379922.2 | NP_001368618.1 | 3 - 4 | 370 | 84 | ||
| NM_001379925.1 | NP_001368620.1 | 3 - 4 | 373 | 84 | ||
| NR_144561.1 | 3 - 4 | 339 | 84 | |||
Set Membership: |
|